Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSRC1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PSRC1 |
---|---|
Concentration | 0.47 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5571
![]() ![]() |
Proteintech Group Inc
271301AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a proline-rich protein. Studies of this gene and the related mouse gene suggest that this gene is regulated by p53 and may participate in p53-mediated growth suppression. The encoded protein may function as a microtubule destabilizing protein that controls spindle dynamics and mitotic progression by recruiting and regulating microtubule depolymerases. At least one genetic variation in this gene has been associated with decreased serum levels of low-density lipoprotein cholesterol. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
PSRC1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q6PGN9 | |
84722 | |
PSRC1 Fusion Protein Ag26039 | |
Primary | |
-20°C | |
PSRC1 |
0.47 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DDA3, FP3214, PSRC1, RP11 297O4.2 | |
PSRC1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title