Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PST1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$162.36 - $492.89
Specifications
| Antigen | PST1 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PST1 Polyclonal specifically detects PST1 in Yeast samples. It is validated for Western Blot, ELISA.Specifications
| PST1 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 851625 | |
| Antibody was raised against a synthetic peptide from the amino terminus of yeast Pst1. The immunogen is located within amino acids 80 - 130 of Pst1. Amino Acid Squence: DSLESITDSLNLQSLT | |
| Primary | |
| 49 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Yeast | |
| Cell wall mannoprotein PST1, D4214, Haze protective factor 2, HPF2, YD9609.09, YDR055W | |
| PST1 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title