Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PST1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP241063 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP241063 0.1 mg
NB396526 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP241063 Supplier Novus Biologicals Supplier No. NBP241063
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PST1 Polyclonal specifically detects PST1 in Yeast samples. It is validated for Western Blot, ELISA.

Specifications

Antigen PST1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA 1 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias Cell wall mannoprotein PST1, D4214, Haze protective factor 2, HPF2, YD9609.09, YDR055W
Gene Symbols PST1
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide from the amino terminus of yeast Pst1. The immunogen is located within amino acids 80 - 130 of Pst1. Amino Acid Squence: DSLESITDSLNLQSLT
Molecular Weight of Antigen 49 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 851625
Target Species Yeast
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.