Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PST1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB355453 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355453 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355453 Supplier Novus Biologicals Supplier No. NBP241063MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PST1 Polyclonal antibody specifically detects PST1 in Yeast samples. It is validated for Western Blot, ELISA

Specifications

Antigen PST1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Cell wall mannoprotein PST1, D4214, Haze protective factor 2, HPF2, YD9609.09, YDR055W
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide from the amino terminus of yeast Pst1. The immunogen is located within amino acids 80 - 130 of Pst1. Amino Acid Squence: DSLESITDSLNLQSLT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 851625
Target Species Yeast
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.