Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSTAIR Antibody (PSTAIR), Novus Biologicals™
SDP

Catalog No. NB12010345 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12010345 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12010345 Supplier Novus Biologicals Supplier No. NB12010345
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSTAIR Monoclonal specifically detects PSTAIR in Human, Mouse, Avian, Plant, Primate, Xenopus samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen PSTAIR
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone PSTAIR
Concentration 7.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500
Gene Alias EGVPSTAIREISLLKE, EGVPSTAIREISLLKE sequence, PSTAIR sequence
Host Species Mouse
Immunogen Synthetic peptide: EGVPSTAIREISLLKE conjugated to BSA.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Test Specificity PSTAIR (PSTAIR)
Target Species Human, Mouse, Avian, Plant, Primate, Xenopus species
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.