Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTBP1 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001310 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001310 Supplier Novus Biologicals Supplier No. NB1001310
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

PTBP1 Polyclonal specifically detects PTBP1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.

Specifications

Antigen PTBP1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1-0.3 ug/ml, Immunohistochemistry 2 - 4 ug/ml, Immunohistochemistry-Paraffin 2-4 ug/ml, Peptide ELISA detection Limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_002810
Gene Alias Heterogeneous nuclear ribonucleoprotein I, heterogeneous nuclear ribonucleoprotein polypeptide I, hnRNP I, HNRNPI, HNRNP-I, HNRPI, MGC10830, MGC8461, polypyrimidine tract binding protein (heterogeneous nuclear ribonucleoproteinI), polypyrimidine tract binding protein 1, polypyrimidine tract-binding protein 1, pPTB, PTB-1, PTB2, PTB3, PTB4, PTB57 kDa RNA-binding protein PPTB-1, PTB-T, RNA-binding protein
Gene Symbols PTBP1
Host Species Goat
Immunogen Peptide with sequence DGIVPDIAVGTKR-C corresponding to N-Terminus according to NP_114367.1, NP_114368.1, NP_787041.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5725
Test Specificity This antibody is expected to recognize all 4 reported isoforms (represented by NP_002810.1; NP_114367.1; NP_114368.1; NP_787041.1).
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.