Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTEN Antibody (217702R), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB295621 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295621 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295621 Supplier Novus Biologicals Supplier No. FAB847RL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PTEN Monoclonal antibody specifically detects PTEN in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, KnockDown

Specifications

Antigen PTEN
Applications Western Blot, Flow Cytometry, KnockDown
Classification Monoclonal
Clone 217702R
Conjugate DyLight 550
Dilution Western Blot, Intracellular Staining by Flow Cytometry, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias BZS, EC 3.1.3.16,10q23del, EC 3.1.3.48, EC 3.1.3.67, GLM2, MMAC1 phosphatase and tensin homolog deleted on chromosome 10, MMAC1MGC11227, Mutated in multiple advanced cancers 1, phosphatase and tensin homologDEC, phosphatidylinositol-34,5-trisphosphate 3-phosphatase and dual-specificityprotein phosphatase PTEN, PTEN1, TEP1MHAM
Host Species Mouse
Immunogen E. coli-derived recombinant human PTEN, Thr2-Val403, Accession # P60484
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cell Biology, Lipid and Metabolism, Metastasis, mTOR Pathway, Phospho Specific, Protein Phosphatase, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5728
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.