Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTER Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP177355 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177355 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177355 Supplier Novus Biologicals Supplier No. NBP177355
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTER Polyclonal specifically detects PTER in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PTER
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. CAH73146
Gene Alias EC 3.1, hPHRP, Parathion hydrolase-related protein, phosphotriesterase related, phosphotriesterase-related protein, resiniferatoxin-binding, phosphotriesterase-related, RPR-1
Gene Symbols PTER
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human PTER. The immunogen is located within amino acids 250 - 300 of PTER. Amino Acid Squence: PDIDMPDDNKRIRRVR
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9317
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.