Learn More
Description
Specifications
Specifications
| Antigen | PTF1A |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | bHLH transcription factor p48, BHLHA29, bHLHa29exocrine pancreas-specific transcription factor p48, Class A basic helix-loop-helix protein 29, p48 DNA-binding subunit of transcription factor PTF1, pancreas specific transcription factor, 1a, pancreas transcription factor 1 subunit alpha, Pancreas-specific transcription factor 1a, PTF1P48, PTF1-p48class II bHLH protein PTF1A |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human PTF1A. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
