Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTF1A Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB105325 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB105325 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB105325 Supplier Novus Biologicals Supplier No. NBP298726AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTF1A Polyclonal specifically detects PTF1A in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PTF1A
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias bHLH transcription factor p48, BHLHA29, bHLHa29exocrine pancreas-specific transcription factor p48, Class A basic helix-loop-helix protein 29, p48 DNA-binding subunit of transcription factor PTF1, pancreas specific transcription factor, 1a, pancreas transcription factor 1 subunit alpha, Pancreas-specific transcription factor 1a, PTF1P48, PTF1-p48class II bHLH protein PTF1A
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human PTF1A.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 256297
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.