Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PTFE-Lined Closed Caps

Catalog No. 14966103
Click to view available options
Neck Finish:
24-414
28-400
33-400
38-400
45-400
53-400
58-400
70-400
83-400
89-400
Color:
White
Product Type:
Closure with Bonded Liner
Solid Top Closure

Use these caps as replacements for our glass environmental sample bottles, jars, jugs and vials.

Thermo Scientific™ PTFE-Lined Closed Caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials. The same high quality PTFE-lined caps used on our Thermo Scientific glass environmental sample containers. The PTFE liner is bonded to the cap so it won't fall out or become separated from the cap.

  • White polypropylene closed top cap with bonded PTFE liner
  • 24-414 cap has a PTFE-faced silicone liner for added cushion and an extra secure seal when used with bottles and VOA vials

Includes:

Cap with bonded liner

Compatible with:

For use with Thermo Scientific glass environmental sample bottles, jars, jugs and vials.

TRUSTED_SUSTAINABILITY

Specifications

Certification Parameters None
Color White
Description Closed top cap
Material Polypropylene
No. per Case 24
Level Unprocessed
Liner Material PTFE
Neck Finish 53-400
Certifications/Compliance Manufactured under an ISO certified quality management system.
Shape Round
Product Type Solid Top Closure
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.