Learn More
Description
Specifications
Specifications
| Antigen | PTGER2/3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 3F12 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 5-10 ug/ml |
| Formulation | PBS with 0.08% Sodium Azide |
| Gene Accession No. | P43116 |
| Gene Alias | EP2, EP3, EP3-I, EP3-II, EP3-III, EP3-IV, PGE receptor EP2 subtype, PGE receptor, EP3 subtype, PGE2 receptor EP2 subtype, PGE2 receptor EP3 subtype, prostaglandin E receptor 2 (subtype EP2), 53kDa, prostaglandin E receptor 3 (subtype EP3), prostaglandin E2 receptor EP2 subtype, prostaglandin E2 receptor EP3 subtype, prostaglandin receptor (PGE-2), Prostanoid EP2 receptor, Prostanoid EP3 receptor |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
