Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTGER2 Antibody (SR1927), Novus Biologicals™
SDP

Catalog No. NB259628 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259628 50 μg
NB258478 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB259628 Supplier Novus Biologicals Supplier No. NBP32175650UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PTGER2 Monoclonal antibody specifically detects PTGER2 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence

Specifications

Antigen PTGER2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone SR1927
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias EP2, PGE receptor EP2 subtype, PGE2 receptor EP2 subtype, prostaglandin E receptor 2 (subtype EP2), 53kD, prostaglandin E receptor 2 (subtype EP2), 53kDa, prostaglandin E2 receptor EP2 subtype, Prostanoid EP2 receptor
Host Species Rabbit
Immunogen A synthesized peptide derived from human PTGER2 (Uniprot #: P43116)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5732
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.