Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTGER3 Antibody (606609) [DyLight 594], Novus Biologicals™
SDP

Catalog No. NB402220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402220 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402220 Supplier Novus Biologicals Supplier No. FAB10243M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PTGER3 Monoclonal antibody specifically detects PTGER3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen PTGER3
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 606609
Conjugate DyLight 594
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EP3EP3e, EP3-I, EP3-II, EP3-III, EP3-IV, MGC141828, MGC141829, MGC27302, PGE receptor EP3 subtype, PGE receptor, EP3 subtype, PGE2 receptor EP3 subtype, PGE2-R, prostaglandin E receotor EP3 subtype 3 isoform, prostaglandin E receptor 3 (subtype EP3), prostaglandin E2 receptor EP3 subtype, prostaglandin receptor (PGE-2), Prostanoid EP3 receptor
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human PTGER3, Met1-Arg390, Accession # P43115
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5733
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.