Learn More
Invitrogen™ PTGES Polyclonal Antibody

Description
PTGES Polyclonal Antibody detects endogenous levels of PTGES protein.
Specifications
Specifications
| Antigen | PTGES |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PTGES |
| Gene Accession No. | O14684, Q9JM51 |
| Gene Alias | 2410099E23Rik; D2Ertd369e; glutathione S-transferase 1-like 1; MGST1L1; MGST1-L1; MGST1-like 1; MGST-IV; Microsomal glutathione S-transferase 1-like 1; microsomal prostaglandin E synthase 1; microsomal prostaglandin E synthase-1; MPGES; MPGES1; mPGES-1; p53-induced apoptosis protein 12; p53-induced gene 12 protein; Pges; PIG12; PP102; PP1294; Prostaglandin E synthase; PTGES; TP53I12; tumor protein p53 inducible protein 12 |
| Gene Symbols | PTGES |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.