Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTGFR Antibody (1014602), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB340098 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340098 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340098 Supplier Novus Biologicals Supplier No. FAB10294MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PTGFR Monoclonal specifically detects PTGFR in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen PTGFR
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 1014602
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FP, MGC120498, MGC46203, PGF receptor, PGF2 alpha receptor, PGF2-alpha receptor, prostaglandin F receptor (FP), prostaglandin F2 alpha receptor, prostaglandin F2-alpha receptor, prostaglandin receptor (2-alpha), Prostanoid FP receptor
Host Species Mouse
Immunogen Synthetic peptide containing human PTGFR RD3 epitope, Accession # P43088
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 5737
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.