Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTH1R/PTHR1 Antibody (003), PerCP, Novus Biologicals™
SDP

Catalog No. NB317002 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB317002 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB317002 Supplier Novus Biologicals Supplier No. NBP290079PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PTH1R/PTHR1 Monoclonal antibody specifically detects PTH1R/PTHR1 in Human samples. It is validated for ELISA

Specifications

Antigen PTH1R/PTHR1
Applications ELISA
Classification Monoclonal
Clone 003
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias MGC138426, parathyroid hormone 1 receptorMGC138452, parathyroid hormone/parathyroid hormone-related peptide receptor, parathyroid hormone/parathyroid hormone-related protein receptor, PFE, PTH/PTHr receptor, PTH/PTHrP type I receptor, PTH1 receptor, PTHRPTHR1parathyroid hormone receptor 1, seven transmembrane helix receptor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human PTH1R/PTHR1 (Uniprot#: Q03431; Met1-Gly188).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, GPCR, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5745
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.