Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTH2R Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7205931 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9258 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9258 Supplier Proteintech Group Inc Supplier No. 141661AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTHR2 is a specific receptor for parathyroid hormone. The activity of this receptor is mediated by g proteins which activate adenylyl cyclase. PTHR2 may be responsible for PTH effects in a number of physiological systems, and may also play a significant role in pancreatic function. PTHR2 presence in neurons indicates that it may function as a neurotransmitter receptor.

Specifications

Antigen PTH2R
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PTH2R
Gene Accession No. P49190, P70555, Q91V95
Gene Alias parathyroid hormone 2 receptor, PTH2 receptor, PTH2R, PTHR2
Gene Symbols PTH2R
Host Species Rabbit
Immunogen PTH2R Fusion Protein Ag5364
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213527, 5746, 81753
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.