Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTIP Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB291763 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291763 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291763 Supplier Novus Biologicals Supplier No. NB100577F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTIP Polyclonal specifically detects PTIP in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen PTIP
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias CAGF28, PACIP1, PAX interacting (with transcription-activation domain) protein 1, PAX transactivation activation domain-interacting protein, PAX transcription activation domain interacting protein 1 like, PAX-interacting protein 1, PAXIP1LCAGF29, protein encoded by CAG trinucleotide repeats, PTIPFLJ41049, TNRC2
Host Species Rabbit
Immunogen Maps to a region between residues 1 and 50 of human PAX transcription activation domain interacting protein 1 using the numbering given in Jowsey et akl., 2004, J. Biol. Chem. 279(53):55562-55569 (GeneID 22976).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 22976
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.