Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTK7/CCK4 Antibody (525222), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB369965 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369965 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369965 Supplier Novus Biologicals Supplier No. FAB4499R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PTK7/CCK4 Monoclonal specifically detects PTK7/CCK4 in Human, Rat samples. It is validated for Immunohistochemistry, Western Blot.

Specifications

Antigen PTK7/CCK4
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 525222
Conjugate Alexa Fluor 647
Dilution Western Blot, Simple Western, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias CCK-4, CCK4Colon carcinoma kinase 4, colon carcinoma kinase-4, Protein-tyrosine kinase 7, Pseudo tyrosine kinase receptor 7, PTK7 protein tyrosine kinase 7, Tyrosine-protein kinase-like 7
Host Species Mouse
Immunogen E. coli-derived recombinant human PTK7/CCK4, Ala31-Ser199, Accession # Q13308
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5754
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.