Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTK7/CCK4 Antibody (OTI2E7), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N273708APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273708APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273708APC7 Supplier Novus Biologicals Supplier No. NBP273708APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PTK7/CCK4 Monoclonal specifically detects PTK7/CCK4 in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen PTK7/CCK4
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2E7
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CCK-4, CCK4Colon carcinoma kinase 4, colon carcinoma kinase-4, Protein-tyrosine kinase 7, Pseudo tyrosine kinase receptor 7, PTK7 protein tyrosine kinase 7, Tyrosine-protein kinase-like 7
Gene Symbols PTK7
Host Species Mouse
Immunogen Full length recombinant protein of human PTK7/CCK4 (NP_690619) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5754.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.