Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTK9 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220001 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220001 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220001 Supplier Novus Biologicals Supplier No. NBP220001
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTK9 Polyclonal specifically detects PTK9 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PTK9
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Alias A6, A6 protein tyrosine kinase, Protein A6, Protein tyrosine kinase 9MGC23788, PTK9 protein tyrosine kinase 9, PTK9MGC41876, twinfilin, actin-binding protein, homolog 1 (Drosophila), twinfilin-1
Gene Symbols TWF1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PTK9. The exact sequence is proprietary.
Molecular Weight of Antigen 29 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5756
Test Specificity Human. Species reactivity based on sequence identity: Mouse (92%), Rat (94%), Bovine (95%), Xenopus tropicalis (80%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.