Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTK9 Antibody (OTI5C12), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N273710A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273710A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273710A700 Supplier Novus Biologicals Supplier No. NBP273710AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PTK9 Monoclonal specifically detects PTK9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.

Specifications

Antigen PTK9
Applications Western Blot, Immunohistochemistry, Immunoassay
Classification Monoclonal
Clone OTI5C12
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Knockout Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias A6, A6 protein tyrosine kinase, Protein A6, Protein tyrosine kinase 9MGC23788, PTK9 protein tyrosine kinase 9, PTK9MGC41876, twinfilin, actin-binding protein, homolog 1 (Drosophila), twinfilin-1
Gene Symbols TWF1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 140-384 of human PTK9 (NP_002813) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5756.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.