Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTOP Antibody (OTI2D5), HRP, Novus Biologicals™
SDP

Catalog No. NBP273711H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273711H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273711H Supplier Novus Biologicals Supplier No. NBP273711H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PTOP Monoclonal specifically detects PTOP in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PTOP
Applications Western Blot
Classification Monoclonal
Clone OTI2D5
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias ALS2CR7, amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 7, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 7 protein, Cell division protein kinase 15, cyclin-dependent kinase 15, EC 2.7.11, EC 2.7.11.22, PFTAIRE protein kinase 2, PFTK2, Serine/threonine-protein kinase ALS2CR7, Serine/threonine-protein kinase PFTAIRE-2
Gene Symbols CDK15
Host Species Mouse
Immunogen Full length recombinant protein of human PTOP (NP_631897) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 65061.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.