Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTP epsilon Antibody, Novus Biologicals™
SDP

Catalog No. NBP162460 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP162460 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP162460 Supplier Novus Biologicals Supplier No. NBP162460
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTP epsilon Polyclonal specifically detects PTP epsilon in Human samples. It is validated for Western Blot.

Specifications

Antigen PTP epsilon
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P23469
Gene Alias DKFZp313F1310, EC 3.1.3.48, FLJ57799, FLJ58245, HPTPE, protein tyrosine phosphatase epsilon, protein tyrosine phosphatase, receptor type, E, protein tyrosine phosphatase, receptor type, epsilon polypeptide, Protein-tyrosine phosphatase epsilon, PTPE, receptor-type tyrosine-protein phosphatase epsilon, R-PTP-EPSILON
Gene Symbols PTPRE
Host Species Rabbit
Immunogen Synthetic peptides corresponding to PTPRE(protein tyrosine phosphatase, receptor type, E) The peptide sequence was selected from the middle region of PTPRE (NP_006495). Peptide sequence VILSMKRGQEYTDYINASFIDGYRQKDYFIATQGPLAHTVEDFWRMIWEW.
Molecular Weight of Antigen 81 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline GPCR, Protein Phosphatase, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 5791
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.