Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTP epsilon Antibody (OTI5F2), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP273713A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273713A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273713A30 Supplier Novus Biologicals Supplier No. NBP273713AF350

Mouse Monoclonal Antibody

PTP epsilon Monoclonal specifically detects PTP epsilon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PTP epsilon
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI5F2
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DKFZp313F1310, EC 3.1.3.48, FLJ57799, FLJ58245, HPTPE, protein tyrosine phosphatase epsilon, protein tyrosine phosphatase, receptor type, E, protein tyrosine phosphatase, receptor type, epsilon polypeptide, Protein-tyrosine phosphatase epsilon, PTPE, receptor-type tyrosine-protein phosphatase epsilon, R-PTP-EPSILON
Gene Symbols PTPRE
Host Species Mouse
Immunogen Full length recombinant protein of human PTP epsilon (NP_006495) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline GPCR, Protein Phosphatase, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 5791.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.