Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTP-MEG2/PTPN9 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB186667 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB186667 200 μg
NB186668 50 μg
NB186666 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. NB186667 Supplier Novus Biologicals Supplier No. NBP312800200UL
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PTP-MEG2/PTPN9 Polyclonal antibody specifically detects PTP-MEG2/PTPN9 in Human samples. It is validated for ELISA, Immunoprecipitation

Specifications

Antigen PTP-MEG2/PTPN9
Applications ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000, Immunoprecipitation 4 to 8 μL/mg of lysate
Formulation 0.2μm filtered solution in PBS without Preservative
Gene Alias EC 3.1.3.48, MEG2, protein tyrosine phosphatase, non-receptor type 9, Protein-tyrosine phosphatase MEG2, PTPase MEG2, PTPase-MEG2, PTPMEG2, tyrosine-protein phosphatase non-receptor type 9
Host Species Rabbit
Immunogen Purified, recombinant Human PTP-MEG2/PTPN9 (NP_002824.1; Ala285-Gln593)
Purification Method Protein A purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5780
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.