Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTP rho/PTPRT Antibody, Novus Biologicals™
SDP

Catalog No. NB10075300 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10075300 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10075300 Supplier Novus Biologicals Supplier No. NB10075300
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

PTP rho/PTPRT Polyclonal specifically detects PTP rho/PTPRT in Human, Rat samples. It is validated for Western Blot.

Specifications

Antigen PTP rho/PTPRT
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:200 - 1:500
Formulation PBS (pH 7.2) with 0.02% Sodium Azide
Gene Accession No. O14522
Gene Alias EC 3.1.3, EC 3.1.3.48, KIAA0283RPTPrho, protein tyrosine phosphatase, receptor type, T, receptor protein tyrosine phosphatase, Receptor-type tyrosine-protein phosphatase rho, receptor-type tyrosine-protein phosphatase T, RPTP-rho, R-PTP-T
Gene Symbols PTPRT
Host Species Chicken
Immunogen The immunogen for this antibody was a sequence from 383a.a. to 484a.a. from PTP rho(a 1444a.a. long protein)
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11122
Test Specificity PTP rho
Target Species Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.