Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTP1B/PTPN1 Antibody (001), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB107315 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB107315 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB107315 Supplier Novus Biologicals Supplier No. NBP289399AF350
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PTP1B/PTPN1 Monoclonal specifically detects PTP1B/PTPN1 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PTP1B/PTPN1
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 1
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, tyrosine-protein phosphatase non-receptor type 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human PTP1B/PTPN1 (NP_002818.1; Glu 2-Asn 321).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 5770
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.