Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTP1B/PTPN1 Antibody (OTI1D10), HRP, Novus Biologicals™
SDP

Catalog No. NBP273729H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273729H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273729H Supplier Novus Biologicals Supplier No. NBP273729H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PTP1B/PTPN1 Monoclonal specifically detects PTP1B/PTPN1 in Human, Rat, Canine, Monkey, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen PTP1B/PTPN1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
Classification Monoclonal
Clone OTI1D10
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated
Formulation PBS with No Preservative
Gene Alias EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, tyrosine-protein phosphatase non-receptor type 1
Gene Symbols PTPN1
Host Species Mouse
Immunogen Full length recombinant protein of human PTP1B/PTPN1 (NP_002818) produced in HEK293 cell.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 5770.0
Target Species Human, Rat, Canine, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.