Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTP4A1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10093524 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093524 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093524 Supplier Novus Biologicals Supplier No. NB10093524
Add to Cart
Add to Cart

Goat Polyclonal Antibody

PTP4A1 Polyclonal specifically detects PTP4A1 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen PTP4A1
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_003454.1
Gene Alias EC 3.1.3.48, HH72, PRL-1DKFZp779M0721, Protein tyrosine phosphatase IVA1, protein tyrosine phosphatase type IVA 1, protein tyrosine phosphatase type IVA protein 1, protein tyrosine phosphatase type IVA, member 1, Protein-tyrosine phosphatase 4a1, Protein-tyrosine phosphatase of regenerating liver 1, PTP(CAAXI), PTPCAAX1PRL1PTP(CAAX1)
Gene Symbols PTP4A1
Host Species Goat
Immunogen Peptide with sequence KDSNGHRNNCCIQ corresponding to C-Terminus according to NP_003454.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7803
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse, Rat and Dog.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.