Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPMT1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB322655 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB322655 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB322655 Supplier Novus Biologicals Supplier No. NBP300056JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTPMT1 Polyclonal antibody specifically detects PTPMT1 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen PTPMT1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias DUSP23, EC 3.1.3.16, EC 3.1.3.48, FLJ46081, MOSPNB4 apoptosis/differentiation related protein, Phosphoinositide lipid phosphatase, PLIPprotein-tyrosine phosphatase mitochondrial 1, protein tyrosine phosphatase, mitochondrial 1, PTEN-like phosphatase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human PTPMT1 (Uniprot#: Q8WUK0-1; Lys28-Thr201)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 114971
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.