Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTPN11 Mouse anti-Human, Mouse, Rat, Clone: 3F8A8, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | PTPN11 |
---|---|
Clone | 3F8A8 |
Concentration | 0.8 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7195
![]() ![]() |
Proteintech Group Inc
667951IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains two tandem Src homology-2 domains, which function as phospho-tyrosine binding domains and mediate the interaction of this PTP with its substrates. This PTP is widely expressed in most tissues and plays a regulatory role in various cell signaling events that are important for a diversity of cell functions, such as mitogenic activation, metabolic control, transcription regulation, and cell migration. Mutations in this gene are a cause of Noo syndrome as well as acute myeloid leukemia.Specifications
PTPN11 | |
0.8 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BPTP3, CFC, NS1, PTP 1D, PTP 2C, PTP2C, PTPN11, SH PTP2, SH PTP3, SHP 2, SHP2, SHPTP2 | |
PTPN11 | |
IgG1 | |
Protein G | |
Antibody |
3F8A8 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P35235, P41499, Q06124 | |
19247, 25622, 5781 | |
PTPN11 Fusion Protein Ag13649 | |
Primary | |
-20°C | |
PTPN11 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title