Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPN12 Recombinant Rabbit Monoclonal Antibody (BL-5-2F8)

Rabbit Recombinant Monoclonal Antibody

$173.10 - $436.00

Specifications

Antigen PTPN12
Clone BL-5-2F8
Concentration 40 μg/mL
Content And Storage 4°C
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIMA544209
View Documents
Invitrogen™
MA544209
100 μL
Each for $436.00
Only null left
Add to Cart
 
PIMA544210
View Documents
Invitrogen™
MA544210
20 μL
Each for $173.10
Only null left
Add to Cart
 
Description

Description

Epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

PTP PEST is an 88 kDa cytosolic protein tyrosine phosphatase which is ubiquitiously expressed in mammalian tissues. PTP PEST exhibits a high specific activity when assayed in vitro with artificial tyrosine-phosphorylated substrates. PTP PEST is subject to regulation via phosphorylation of ser 39 and is catalyzed by both protein kinase C and protein kinase A.
Specifications

Specifications

PTPN12
40 μg/mL
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6
Q05209
5782
Synthetic peptide representing a region between residues 499 and 780 of human Protein Tyrosine Phosphatase, non-receptor type 12.
Antibody
BL-5-2F8
4°C
Recombinant Monoclonal
Liquid
IgG
Human
PTPN12
MPTP-PEST; P19-PTP; pest; protein tyrosine phosphatase non-receptor type 12; protein tyrosine phosphatase, non-receptor type 12; protein-tyrosine phosphatase G1; Protein-tyrosine phosphatase P19; PTPG1; PTPN12; PTP-P19; PTP-PEST; RKPTP; tcag7.1075; Tyrosine-protein phosphatase non-receptor type 12
PTPN12
Primary
Purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.