Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN13/PTPL1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056140 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056140 0.05 mL
NB10056140T 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056140 Supplier Novus Biologicals Supplier No. NB10056140
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTPN13/PTPL1 Polyclonal specifically detects PTPN13/PTPL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen PTPN13/PTPL1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000
Formulation Whole antisera with 0.02% Sodium Azide
Gene Accession No. Q12923
Gene Alias hPTPE1, PNP1, protein tyrosine phosphatase, non-receptor type 13 (APO-1/CD95 (Fas)-associatedphosphatase), protein-tyrosine phosphatase 1, Fas-associated, PTP1EDKFZp686J1497, PTP-BASEC 3.1.3.48, PTP-E1, PTPL1nonreceptor type 13, tyrosine-protein phosphatase non-receptor type 13
Gene Symbols PTPN13
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1760-1775 (EPTRQENWTPLKNDLE) of human FAP-1 was used as immunogen; GenBank no. NP_542414.1. This peptide sequence is conserved in multiple isoforms of FAP-1. These FAP-1 isoforms are of varying amino acid (aa) lengths and include: (1) NP_542414.1 = 2485 aa, FAP-1 isoform 1. (2) BAD92141.1 = 2434 aa, FAP-1 isoform 2. (3) NP_542415.1 = 2294 aa, FAP-1 isoform 3. (4) NP_542416.1 = 2490 aa, FAP-1 isoform 4. (5) AAH39610.1 = 604 aa, FAP-1. (6) AAF63474.1 = 499 aa, FAP-1. For additional information on FAP-1 isoforms, users are encouraged to use the NCBI data bases such as AceView (http://www.ncbi.nlm.nih.gov/IEB/Research/Acembly/) or BLAST (http://www.ncbi.nlm.nih.gov/BLAST/).
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 5783
Test Specificity FAP-1 typically migrates at ∼270 kDa on western blots. Thus for SDS-PAGE and western blot, the researcher should use techniques optimized for the transfer of proteins with high molecular weights. The actual molecular weight of FAP-1 observed depends on the isoform expression.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.