Learn More
Description
Specifications
Specifications
| Antigen | PTPN23 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg |
| Formulation | Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide |
| Gene Accession No. | NP_056281.1 |
| Gene Alias | EC 3.1.3.48, HDPTP, HD-PTPDKFZp564F0923, His domain-containing protein tyrosine phosphatase, His-domain protein tyrosine phosphatase, KIAA1471DKFZP564F0923, Protein tyrosine phosphatase TD14, protein tyrosine phosphatase, non-receptor type 23, PTP-TD14, tyrosine-protein phosphatase non-receptor type 23 |
| Gene Symbols | PTPN23 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
