Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB001666 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001666 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001666 Supplier Novus Biologicals Supplier No. NBP278797

Rabbit Polyclonal Antibody

PTPN3 Polyclonal specifically detects PTPN3 in Human samples. It is validated for Western Blot.

Specifications

Antigen PTPN3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation TBS containing 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. P26045
Gene Alias DKFZp686N0569, EC 3.1.3.48, protein tyrosin phosphatase H1, protein tyrosine phosphatase, non-receptor type 3, Protein-tyrosine phosphatase H1, PTP-H1, PTPH1cytoskeletal-associated protein tyrosine phosphatase, tyrosine phosphatase, tyrosine-protein phosphatase non-receptor type 3
Gene Symbols PTPN3
Host Species Rabbit
Immunogen between 175 and 225
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 5774
Target Species Human
Content And Storage Store at 2-8°C/ 1 year from date of receipt
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.