Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTPRB Monoclonal Antibody (122.2)
Mouse Monoclonal Antibody
$536.00 - $818.50
Specifications
Antigen | PTPRB |
---|---|
Clone | 122.2 |
Concentration | 1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-674-947
|
Invitrogen™
MA15869 |
50 μL |
Each for $536.00
|
|
|||||
01-674-948
|
Invitrogen™
MA15870 |
100 μL |
Each for $818.50
|
|
|||||
Description
Predicted molecular weight: 224 kDa.
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an extracellular domain, a single transmembrane segment and one intracytoplasmic catalytic domain, thus belongs to receptor type PTP. The extracellular region of this PTP is composed of multiple fibronectin type_III repeats, which was shown to interact with neuronal receptor and cell adhesion molecules, such as contactin and tenascin C. This protein was also found to interact with sodium channels, and thus may regulate sodium channels by altering tyrosine phosphorylation status. The functions of the interaction partners of this protein implicate the roles of this PTP in cell adhesion, neurite growth, and neuronal differentiation. Alternate transcript variants encoding different isoforms have been found for this gene.Specifications
PTPRB | |
1 mg/mL | |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 0.1% sodium azide | |
0 | |
314843 | |
Embryonic rat brain proteoglycans. | |
Antibody |
122.2 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Monoclonal | |
Liquid | |
IgM | |
Rat | |
PTPRB | |
3230402H02Rik; C130094E24; HPTPB; HPTP-BETA; protein tyrosine phosphatase, receptor type B; protein tyrosine phosphatase, receptor type, B; protein tyrosine phosphatase, receptor type, beta polypeptide; protein-tyrosine phosphatase beta; PTPB; Ptprb; Ptpz; receptor-type tyrosine-protein phosphatase beta; Rptpb; R-PTP-BETA; vascular endothelial protein tyrosine phosphatase; VEPTP; VE-PTP | |
PTPRB | |
Primary | |
Size-exclusion - Dialysis |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title