Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPRB Mouse anti-Human, Clone: 2-A2, Novus Biologicals™
SDP

Catalog No. NBP267087 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267087 Supplier Novus Biologicals Supplier No. NBP267087

Mouse Monoclonal Antibody

PTPRB Monoclonal specifically detects PTPRB in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PTPRB
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 2-A2
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Formulation PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias DKFZp686E2262, DKFZp686H15164, EC 3.1.3.48, FLJ44133, HPTPB, HPTP-BETA, MGC142023, MGC59935, protein tyrosine phosphatase, receptor type, B, protein tyrosine phosphatase, receptor type, beta polypeptide, Protein-tyrosine phosphatase beta, PTPB, receptor-type tyrosine-protein phosphatase beta, R-PTP-BETA, Vascular endothelial protein tyrosine phosphatase, VEPTP, VE-PTP
Gene Symbols PTPRB
Host Species Mouse
Immunogen Recombinant protein within Human PTPRB aa 1,018-1,161/ 1,997. (SwissProt: P23467 Human)
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 5787
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.