Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPRD Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB165109 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165109 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB165109 Supplier Novus Biologicals Supplier No. NBP281772F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PTPRD Polyclonal antibody specifically detects PTPRD in Human samples. It is validated for ELISA

Specifications

Antigen PTPRD
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias HPTPDELTA, HPTPMGC119752, MGC119750, MGC119751, protein tyrosine phosphatase, receptor type, D, Protein-tyrosine phosphatase delta, PTPDMGC119753, receptor type, delta polypeptide, receptor-type tyrosine-protein phosphatase delta
Host Species Rabbit
Immunogen PTPRD antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human PTPRD. The immunogen is located within the first 50 amino acids of PTPRD. Amino Acid Squence: RTDAETPPRFTRTPVDQT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5789
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.