Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPRD Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB342459 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342459 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342459 Supplier Novus Biologicals Supplier No. NBP281772MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTPRD Polyclonal antibody specifically detects PTPRD in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen PTPRD
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias HPTPDELTA, HPTPMGC119752, MGC119750, MGC119751, protein tyrosine phosphatase, receptor type, D, Protein-tyrosine phosphatase delta, PTPDMGC119753, receptor type, delta polypeptide, receptor-type tyrosine-protein phosphatase delta
Host Species Rabbit
Immunogen PTPRD antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human PTPRD. The immunogen is located within the first 50 amino acids of PTPRD. Amino Acid Squence: RTDAETPPRFTRTPVDQT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5789
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.