Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PU.1/Spi-1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227163SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227163SS 0.025 mg
NBP227163 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227163SS Supplier Novus Biologicals Supplier No. NBP227163SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PU.1/Spi-1 Polyclonal specifically detects PU.1/Spi-1 in Human, Mouse, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen PU.1/Spi-1
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 4-6ug/ml, Flow Cytometry 1.25 ug/10^6 cells
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. P17947
Gene Alias hematopoietic transcription factor PU.1, OF, PU.1, SFPI131 kDa transforming protein, SPI-1, SPI-1 proto-oncogene, SPI-A, spleen focus forming virus (SFFV) proviral integration oncogene spi1,31 kDa-transforming protein, transcription factor PU.1
Gene Symbols SPI1
Host Species Rabbit
Immunogen A portion of amino acid 20-70 of human PU.1 was used as the immunogen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6688
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, chimpanzee, monkey, dog, panda, horse, cow, and wild pig, 92% homologous in rat, 89% homologous in opossum,77% homologous in zebra finch and clearnose skate, and 71% homologous in chicken and dwarf caiman.
Target Species Human, Mouse, Bovine, Canine, Equine, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.