Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PU.1/Spi-1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $426.00

Specifications

Antigen PU.1/Spi-1
Concentration 0.5 mg/mL
Dilution Western Blot 4-6ug/ml, Flow Cytometry 1.25 ug/10^6 cells
Applications Western Blot, Flow Cytometry
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227163SS
SDP
View Documents
Novus Biologicals
NBP227163SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP227163
SDP
View Documents
Novus Biologicals
NBP227163
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

PU.1/Spi-1 Polyclonal specifically detects PU.1/Spi-1 in Human, Mouse, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

PU.1/Spi-1
Western Blot 4-6ug/ml, Flow Cytometry 1.25 ug/10^6 cells
Polyclonal
Purified
RUO
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
hematopoietic transcription factor PU.1, OF, PU.1, SFPI131 kDa transforming protein, SPI-1, SPI-1 proto-oncogene, SPI-A, spleen focus forming virus (SFFV) proviral integration oncogene spi1,31 kDa-transforming protein, transcription factor PU.1
SPI1
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in human, mouse, chimpanzee, monkey, dog, panda, horse, cow, and wild pig, 92% homologous in rat, 89% homologous in opossum,77% homologous in zebra finch and clearnose skate, and 71% homologous in chicken and dwarf caiman.
0.5 mg/mL
Western Blot, Flow Cytometry
Unconjugated
Rabbit
Human, Mouse, Bovine, Canine, Equine, Primate
P17947
6688
A portion of amino acid 20-70 of human PU.1 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.