Learn More
PU.1/Spi-1 Antibody (PU1/2146), Janelia Fluor™ 646, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | PU.1/Spi-1 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | PU1/2146 |
| Conjugate | Janelia Fluor 646 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | hematopoietic transcription factor PU.1, OF, PU.1, SFPI131 kDa transforming protein, SPI-1, SPI-1 proto-oncogene, SPI-A, spleen focus forming virus (SFFV) proviral integration oncogene spi1,31 kDa-transforming protein, transcription factor PU.1 |
| Gene Symbols | SPI1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.