Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PU.1/Spi-1 His Protein
SDP

Catalog No. NBP25197625 Shop All R&D Systems Products
Click to view available options
Quantity:
0.05 mg
0.25 mg

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-271 of Human PU.1/Spi-1 The Recombinant Human PU.1/Spi-1 Protein is derived from E. coli. The Recombinant Human PU.1/Spi-1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl(pH 8.0) containing 10% glycerol
Gene ID (Entrez) 6688
Molecular Weight (g/mol) TMW: 33.6kDa
Quantity 0.25 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias hematopoietic transcription factor PU.1, OF, PU.1, SFPI131 kDa transforming protein, SPI-1, SPI-1 proto-oncogene, SPI-A, spleen focus forming virus (SFFV) proviral integration oncogene spi1,31 kDa-transforming protein, transcription factor PU.1
Common Name PU.1/Spi-1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.