Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PUF60 Antibody, Novus Biologicals™
SDP

Catalog No. NBP149906 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149906 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149906 Supplier Novus Biologicals Supplier No. NBP149906
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PUF60 Polyclonal specifically detects PUF60 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation (Negative).

Specifications

Antigen PUF60
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunoprecipitation (Negative)
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Alias FBP interacting repressor, FBP-interacting repressor, FIRpoly-U binding splicing factor PUF60, FLJ31379, FUSE-binding protein-interacting repressor, poly(U)-binding-splicing factor PUF60, poly-U binding splicing factor 60KDa, Ro ribonucleoprotein binding protein 1, Ro ribonucleoprotein-binding protein 1, Ro-binding protein 1, roBP1, ROBPI, siah binding protein 1,60 kDa poly(U)-binding-splicing factor, Siah-binding protein 1, siah-BP1, SIAHBP1pyrimidine tract binding splicing factor
Gene Symbols PUF60
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 509 and 559 of human Poly-U Binding Splicing Factor 60KDa using the numbering given in entry NP_510965.1 (GeneID 22827).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22827
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with X. tropicalis, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Green anole, White-tufted-ear marmosetand Naked mole rat.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.