Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PUM1 Antibody, Novus Biologicals™
SDP

Catalog No. NB100259 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100259 Supplier Novus Biologicals Supplier No. NB100259
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 5 publications

PUM1 Polyclonal specifically detects PUM1 in Human, Mouse samples. It is validated for Gel Super Shift Assays, Gel Supershift, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot.

Specifications

Antigen PUM1
Applications Gel Supershift, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1-10 μg/mg lysate, Gel Super Shift Assays 1:1-1:100
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. Q14671
Gene Alias HSPUM, KIAA0099PUML1, PUMH, PUMH1pumilio homolog 1, pumilio (Drosophila) homolog 1, pumilio homolog 1 (Drosophila), Pumilio-1
Gene Symbols PUM1
Host Species Goat
Immunogen The immunogen recognized by this antibody maps to a region between residues 225 and 275 of human Pumilio 1 using the numbering given in entry NP_055491.1 (GeneID 9698).
Molecular Weight of Antigen 126 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9698
Test Specificity This is specific for human Pumilio. Reactivity with Drosophila is likely owing to complete identity between Pumilio 1 from these species and the peptide used for immunogen. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Tasmanian devil, White-tufted-ear marmoset, Crab-eating macaque, African elephant, Naked mole rat and Northern white-cheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.