Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PUM2 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB363090 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363090 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363090 Supplier Novus Biologicals Supplier No. NB100387UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PUM2 Polyclonal specifically detects PUM2 in Human, Mouse, Rat samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen PUM2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias FLJ36528, KIAA0235PUMH2PUML2, MGC138251, MGC138253, pumilio (Drosphila) homolog 2, pumilio homolog 2, pumilio homolog 2 (Drosophila), Pumilio-2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 100 and 150 of human Pumilio 2 using the numbering given in entry NP_056132.1 (GeneID 23369).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 23369
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.