Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PUMA Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB344539 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344539 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344539 Supplier Novus Biologicals Supplier No. NB500230MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PUMA Polyclonal antibody specifically detects PUMA in Human samples. It is validated for Western Blot

Specifications

Antigen PUMA
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias BCL2 binding component 3, bcl-2-binding component 3, JFY1, JFY-1, p53 up-regulated modulator of apoptosis, PUMAFLJ42994
Host Species Rabbit
Immunogen Reacts with a 17 residue sequence [EQHLESPVPSAPGALAG] found in the exon-3-encoded region in the human PUMA-alpha and PUMA-beta forms of the protein. Recognizes both forms of the protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cellular Markers, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mitochondrial Markers, Protein Phosphatase, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 27113
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.