Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PUMA Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB357707 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357707 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357707 Supplier Novus Biologicals Supplier No. NBP176639MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PUMA Polyclonal antibody specifically detects PUMA in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen PUMA
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias BCL2 binding component 3, bcl-2-binding component 3, JFY1, JFY-1, p53 up-regulated modulator of apoptosis, PUMAFLJ42994
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human PUMA isoform 1. The immunogen is located within the first 50 amino acids of PUMA. Amino Acid Squence: ARARQEGSSPEPVEG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cellular Markers, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mitochondrial Markers, Protein Phosphatase, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 27113
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.